Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Fmoc-L-phenylalanine ,35661-40-6,MFCD00037128,25G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 0244325G
Molecular Formula C24H21NO4, Purity Limit ≥ 99.7% (Chiral HPLC), Molecular Weight 387.40, Melting Point 180-187 ºC, Storage Temp: Store at 0-8 °C, Synonyms: Fmoc-L-Phe-OH
Catalog No. 50-495-644