Learn More
Medchemexpress LLC Fmoc-Pra-OH | 198561-07-8 | 99.76% | 335.36 | 25 G

Supplier: Medchemexpress LLC 25GHYW011210
Fmoc-Pra-OH is a Glycine derivative and a click chemistry reagent. It contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. Amino acid derivatives like Fmoc-Pra-OH are used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage.
- For research use only
- Solid appearance
- White to off-white color
- Store powder at -20°C for 3 years, 4°C for 2 years
- Store in solvent at -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.