Learn More
Chem-Impex International, Inc.Supplier Diversity Partner Fmoc-O-tert-butyl-L-threonine,71989-35-0,MFCD00077075,100G DFS Item
Manufacturer:  Chem-Impex International, Inc.Supplier Diversity Partner 02460100G
Molecular Formula C23H27NO5; Purity Limit ≥ 99.7% (HPLC, Chiral purity); Molecular Weight 397.50; Melting Point 130-135 ºC; Storage Temp: Store at 0-8 °C; Synonyms: Fmoc-L-Thr(tBu)-OH
Catalog No. 50-495-711