Learn More
Bachem Fmoc-Trp(Boc)-OH
SDP
Supplier:  Bachem 4017674.0005
Encompass_Preferred
Fmoc-Trp(Boc)-OH, 143824-78-6, C31H30N2O6, Mr 526.59, 5g. Sold under license to US Pat. 5,300,651 and EP 0 516 070. The use of this protected tryptophan derivative in Fmoc solid-phase peptide synthesis resulted in an improved yield of the correct product by suppressing side reactions of tryptophan.
Catalog No. 50-312-740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.