Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Val-Cys Psi Dmp.H pro -OH, Advanced ChemTech

Catalog No. p-200022997
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 1926163-08-7
Molecular Formula C33H36N2O7S
Molecular Weight (g/mol) 604.72
InChI Key HYOCLBUVPYODEB-UHFFFAOYSA-N
IUPAC Name 2-(2,4-dimethoxyphenyl)-4-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanamido]thiolane-3-carboxylic acid
SMILES COC1=CC(OC)=C(C=C1)C1SCC(NC(=O)C(NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(C)C)C1C(O)=O

Specifications

Color White/Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Fmoc-Val-Cys Psi Dmp.H pro -OH
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.