Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMRFamide Rabbit anti-Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$153.60 - $357.12

Specifications

Antigen FMRFamide
Concentration Conc. Not Determined
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0119 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA20002150UL
150 μL
Each for $357.12
Add to cart
 
50-210-0120 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA2000250UL
50 μL
Each for $153.60
Add to cart
 
Description

Description

FMRFamide Polyclonal antibody specifically detects FMRFamide in Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

FMRFamide
Immunohistochemistry
Unconjugated
Rabbit
Rat
FMRF-NH2
Polyclonal
-20° C, Avoid Freeze/Thaw Cycles
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
IgG
Primary
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit