Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FN1 Mouse anti-Human, Clone: 2F4, Abnova™

Mouse monoclonal antibody raised against partial recombinant human FN1.

Manufacturer:  Abnova Corporation MAB12994

Catalog No. 89-934-155


Add to cart

Description

Description

This gene encodes fibronectin, a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. The gene has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some variants has not been determined. [provided by RefSeq]

Specifications

Specifications

FN1
2F4
Mouse monoclonal antibody raised against partial recombinant human FN1.
In ascites (0.03% sodium azide).
CIG/DKFZp686F10164/DKFZp686H0342/DKFZp686I1370/DKFZp686O13149/ED-B/FINC/FN/FNZ/GFND/GFND2/LETS/MSF
Mouse
IgG1
RUO
Primary
2335
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
FN1
Recombinant protein corresponding to amino acids 1965-2176 of human FN1.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit