Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FNDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556603
Description
Immunogen sequence: ECYAEEDEFS GLETDTAVPT EEAYVIYDED YEFETSRPPT TTEPSTTATT PRVIPEEGAI SSFPEEEFDL AGRKRFVAPY VTYLNKDPSA PCSLTDALD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.

Specifications
FNDC1 | |
Polyclonal | |
Unconjugated | |
FNDC1 | |
1110027O12Rik; 1110051A18Rik; Activation-associated cDNA protein; activator of G-protein signaling 8; AGS8; bA243O10.1; dJ322A24.1; expressed in synovial lining protein; fibronectin type III domain containing 1; fibronectin type III domain containing 2; fibronectin type III domain-containing protein 1; FNDC1; FNDC2; KIAA1866; MEL4B3; mKIAA1866 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4ZHG4 | |
FNDC1 | |
Recombinant protein corresponding to Human FNDC1. Recombinant protein control fragment (Product #RP-104190). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction