Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC3A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA5140045

Encompass

Catalog No. PIPA5140045


Add to Cart
This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VNWEVPLSNG TDVTEYRLEW GGVEGSMQIC YCGPGLSYEI KGLSPATTYY CRVQALSVVG AGPFSEVVAC VTPPSVPGIV TCLQEISDDE IENPHYSPST CLAISWEKPC DHGSEILAYS IDFGDKQSLT VGKV

FNDC3A Polyclonal antibody specifically detects FNDC3A in Human samples. It is validated for Immunocytochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FNDC3A
Polyclonal
Unconjugated
FNDC3A
1700094E19Rik; bA203I16.1; bA203I16.5; D14Ertd453e; F730017H24Rik; fibronectin domain-containing protein 3a; fibronectin type III domain containing 3a; fibronectin type-III domain-containing protein 3A; FNDC3; fndc3a; HUGO; Human gene expressed in odontoblasts; KIAA0970; sys; wu:fk65b04; zmp:0000001126
Rabbit
Antigen affinity chromatography
RUO
22862
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
0.05 μg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y2H6
FNDC3A
Recombinant protein corresponding to Human FNDC3A
100 μL
Primary
Human
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit