Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FNDC8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17965720UL

 View more versions of this product

Catalog No. NBP17965720


Add to cart

Description

Description

FNDC8 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

FNDC8
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
FNDC8
Synthetic peptide directed towards the N terminal of human FNDC8The immunogen for this antibody is FNDC8. Peptide sequence MASEALHQVGDGEEAVLKKENFNMMNALDQLPKPFSNPKSMNRTVTTKGL.
36 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_060029
DKFZp434H2215, fibronectin type III domain containing 8, fibronectin type III domain-containing protein 8
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
54752
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit