Learn More
FNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 198471AP
Catalog No. 50-173-2571
Add to cart
Description
Description
FNIP1 encodes a protein that binds to the tumor suppressor protein folliculin and to AMP-activated protein kinase (AMPK). The encoded protein participates in the regulation of cellular metabolism and nutrient sensing by modulating the AMPK and target of rapamycin signaling pathways. This gene has a closely related paralog that encodes a protein with similar binding activities. Both related proteins also associate with the molecular chaperone heat shock protein-90 (Hsp90) and negatively regulate its ATPase activity and facilitate its association with folliculin. Diseases associated with FNIP1 include Birt-Hogg-Dube Syndrome and Pleural Disease.
Specifications
Specifications
FNIP1
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FD7, Q8TF40
FNIP1, KIAA1961
Rabbit
IgG
150 μL
-20°C
Primary
100362347, 216742, 96459
Immunohistochemistry (Paraffin)
Unconjugated
FNIP1
Liquid
FNIP1
FNIP1 Fusion Protein Ag13899
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat