Learn More
3M Company Foam Fast 74 Spray Adhesive
3M Company Foam Fast 74 Spray Adhesive is designed specifically for foams and fabrics
Manufacturer:  3M Company 7000023925
Catalog No. 19-070-909
Add to cart
Description
Description
  • Dispenses with a lace-spray pattern optimized for foam bonding
  • Provides professional, industrial strength for demanding applications
  • Offers a fast, aggressive tack for a quick assembly of high-stress bonds
  • Allows for a bond time of 15 seconds to 30 minutes
  • Fast-drying
  • Bonds foam and fabric to a wide range of substrates
  • Non-yellowing formula
  • Net Wt 16.9 oz,
  • 3M Catalog# 74
Specifications
Specifications
Safety
Adhesives and Sealants
Large Surface Lamination Assembly
Orange
709.77 ml
12