Learn More
BioworldSupplier Diversity Partner Foam Free, 50 g DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220400311
FoamFree™ Specifications:Foam Free, free-flowing dry powder, has been specifically developed for controlling foam in various biotechnology applications. Prevents foaming during sonication or homogenization of cells and tissues for protein extraction, in hybridization solutions during rocking and shaking, and during fermentation of bacterial or yeast cultures. Foam Free is inert, non-toxic, and non-metabolizable; it does not interfere with enzyme activity or assays.
Catalog No. 50-253-029