Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Foamtec MiraWIPE™ Wipers
Suitable for disinfection of vacuum chambers, medical devices and cleanrooms
Supplier: Foamtec HT5790S25
This item is not returnable.
View return policy
Description
MiraWIPE™ wipers feature a non-symmetrical micro fiber structure allows them to scrub microbial biofilms and sporicidal residues.
- Made of continuous filament micro denier, super soft polyester/nylon textile
- High absorbency and contamination pickup
- Abrasion resistant, no-run, no-stretch fabric to minimize In-Use Particles from the face of the wiper
- Available in configurations to meet Class 1-1000 applications
Specifications
MiraWIPE™ Wipes | |
22.86 cm | |
22.86 cm | |
ISO Class 4 | |
9 x 9 in. (22.86 x 22.86 cm) | |
25 Wipers/Bag |
9 in. | |
9 in. | |
Polyester | |
Extra wide sealed | |
For removing stubbornly adhered particles and stains in pharmaceutical and micro electronics cleaning applications, ISO Class 10 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction