Foamtec PharmaMOP™ Cleanroom Mop Heads, Sterile

For cleaning and disinfection of rough surfaces and high soil areas

Manufacturer: Foamtec FS856ENST

 View more versions of this product

Catalog No. 19-304-823

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ISO 9001:2000
    For Use With (Application) For cleaning and disinfection of rough surfaces and high soil areas, steam sterilization cycle
    Weight (English) 1.45 lb.
    Material Polyester
    Product Type EzGLIDE polyester mop head with 0.125 in. foam core
    Sterile Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit