Foamtec PharmaMOP™ Cleanroom Mops
For critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease
Manufacturer: Foamtec FT414D
 View more versions of this product
Catalog No. 19-304-762
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) Critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease
    Length (English) 14 in.
    Includes Locking head frame, lever lock on telescoping handle
    Certifications/Compliance ISO 9001:2000, ISO 13485:2003
    Product Type Cleanroom Mop
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback