Foamtec PolyCLEAN™ Wipers

Lightweight, knitted-wiper constructed and processed to meet clean room requirements

Manufacturer: Foamtec HT5290S50

 View more versions of this product

Catalog No. 19-304-710

  Check Availability
Add to Cart

    Specifications

    Color White
    Adsorbent Type 3.2
    Basis Weight 131
    Construction Continuous filament Fiber
    Dimensions (L x W) 9 x 9 in. (22.86 x 22.86cm)
    Edges Wide sealed
    Material Knit Polyester Fabric
    Width (English) 9 in.
    Width (Metric) 22.86cm
    For Use With (Application) Ideal for particle sensitive wiping applications
    Length (English) 9 in.
    Length (Metric) 22.86cm
    Product Type Polyclean™ cleanroom wipes

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit