Foamtec UltraMOP™ III Mop Heads
For cleaning and disinfecting ISO Class 1 clean rooms
Manufacturer: Foamtec FS704EN
 View more versions of this product
Catalog No. 19-304-603
  Check Availability
Add to Cart
    Specifications
    Material Polyurethane Foam
    Product Type Microfiber lamination with modified T
    Sterile No
    For Use With (Application) Cleaning and disinfecting ISO Class 1 cleanrooms
    Length (English) 12 in.
    Width (English) 4.12 in.
    Bump-Proof™ Wrap-Around design ensures that the leading and trailing edges do not generate particles resulting from abrasion damage. The fabric layer wraps over both the leading and trailing edges improving cleanliness and durability
    • Made of snag-resistant fabrics laminated to a super, fine pore urethane foam
    • Mop heads are available in microfiber laminated versions
  • Description & Specifications
Related Products
Product Content Feedback