Foamtec UltraMOP™ IV Mop Heads, Sterile
Bump-proof™ wrap-around design for cleaning and disinfecting ISO Class 1 clean rooms
Manufacturer: Foamtec FS806ST181
 View all options for this product
Catalog No. 19-304-831
  Check Availability
Add to Cart
    Autoclavable Yes
    Material Polyester Fabric, Urethane Foam
    Product Type Mop Head
    Sterile Yes
    For Use With (Application) For cleaning and disinfecting ISO Class 1 cleanrooms
    Length (English) 18 in.
    • Durable snag resistant fabric construction
    • With additional abrasive mesh surface edge
    • Eliminates rollover, abrasion-related particles
    • Offers smooth, skip-free motion enhancing disinfectant laydown
    • Compatible with gamma, autoclave and ETO sterilization methods
    • Lightweight slide-on design helps for adhering mop head to Slimline™ frame
    • Available in microfiber and polyester laminated versions for stubborn contamination like hardened biofilms and disinfectant residue
  • Description & Specifications
Related Products
Product Content Feedback