Learn More
Foamtec UltraMOP™ IV Mop Heads, Sterile
Bump-proof™ wrap-around design for cleaning and disinfecting ISO Class 1 clean rooms
Supplier: Foamtec FS806ST141
Description
- Durable snag resistant fabric construction
- With additional abrasive mesh surface edge
- Eliminates rollover, abrasion-related particles
- Offers smooth, skip-free motion enhancing disinfectant laydown
- Compatible with gamma, autoclave and ETO sterilization methods
- Lightweight slide-on design helps for adhering mop head to Slimline™ frame
- Available in microfiber and polyester laminated versions for stubborn contamination like hardened biofilms and disinfectant residue