Foamtec UltraSOLV™ ScrubDISKs
Manufacturer: Foamtec HT4580DC31
 View more versions of this product
Catalog No. 19-120-2192
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) ErgoScrub hand tool
    For Use With (Application) Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear
    Adhesive Pressure sensitive adhesive
    Construction Fiber-free
    Diameter (English) 3.5 in.
    Diameter (Metric) 8.89cm
    Disposable Yes
    Grit Rating 800
    Includes With pressure-sensitive adhesive or hook and loop fastener
    Material Diamond
    Product Type Cleaning disk
  • Description & Specifications
Related Products
Product Content Feedback