Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
Foamtec UltraSOLV Wipers
For cleaning critical tools and assemblies
Manufacturer: Foamtec HT4791
Description
Static dissipative and tribo charging properties allow for safe cleaning of static sensitive assemblies, such as wafer transfer chambers in vacuum tools
- Highly absorbent
- Class 10 Wiper constructed from naturally reticulated, hydrophilic, polyurethane foam
- Abrasion resistance
- Low levels of particle generation, ionics, organics, lot-to-lot consistency
Specifications
9 in. | |
9 x 9 in. (22.86 x 22.86 cm) | |
9 in. | |
Polyurethane, Foam | |
For safe cleaning of static sensitive assemblies, such as wafer transfer chambers in vacuum tools |
22.86 cm | |
For cleaning critical tools and assemblies, Class 10 | |
22.86 cm | |
Ultrasolv wipes | |
0.276 cm |