Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marvel™ Focus Desk™ XT with Furniture Casters

Adjustable desk height from 28 - 42 in.tall in 3 sec. without assistance, tools and electricity

Manufacturer:  Marvel™ FSDXT3220BSCMST

 View more versions of this product

Catalog No. 08-101-195

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Adjustable height from 28 - 42 in. with FeatherTouch™ Lift System
  • FeatherTouch Lift System allows to quickly and easily adjust shelf height with one hand
  • Height adjustment lever is spring assisted
  • Upper and lower desk supports are constructed of 12 gauge steel
  • Each 2 in. furniture caster has brake
    • Certifications

      Greenguard, ISO 9001-2008 and UL Certified

      Warranty and Services

      1 year

Specifications

Specifications

28 - 42 in.
71.1 - 106.6cm
Box, side shelf and casters
Sit/Stand Desk
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit