Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marvel™ Focus Desk™ XT with Furniture Casters

Adjustable desk height from 28 - 42 in.tall in 3 sec. without assistance, tools and electricity

Specifications

Product Type Sit/Stand Desk
View More Specs

Products 4
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
08-101-192
View Documents
Marvel™
FSDXT3220CMST
Casters This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
08-101-193
View Documents
Marvel™
FSDXT3220BCKMST
Book box and casters This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
08-101-194
View Documents
Marvel™
FSDXT3220SCMST
Side shelf and casters This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
08-101-195
View Documents
Marvel™
FSDXT3220BSCMST
Box, side shelf and casters This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Adjustable height from 28 - 42 in. with FeatherTouch™ Lift System
  • FeatherTouch Lift System allows to quickly and easily adjust shelf height with one hand
  • Height adjustment lever is spring assisted
  • Upper and lower desk supports are constructed of 12 gauge steel
  • Each 2 in. furniture caster has brake
    • Certifications

      Greenguard, ISO 9001-2008 and UL Certified

      Warranty and Services

      1 year

Specifications

Specifications

Sit/Stand Desk
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit