Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nasco Focused Assessment with Sonography for Trauma

Supplier: Nasco BPFAST1800
Description
- Fully Imageable Combination FAST, TTE, and Pericardiocentesis Ultrasound Model
- Extremely realistic combination Transthoracic Echo (TTE), Pericardiocentesis, and Focused Assessment with Sonography for Trauma (FAST) exam ultrasound model
- Adjust internal bleeding states around the liver, spleen, heart, and bladder, simulating a wide range of pathological scenarios
- Fully imageable upper and lower torso; contains the liver, gallbladder, kidneys, spleen, heart, pericardial fluid, lungs, ribs, bowel, bladder, stomach, and skeleton
- Model extends from head to mid-thigh
- Heart contains accurate anatomy including left and right ventricle, left and right atria, mitral valve, tricuspid valve, aortic valve, pulmonic valve, left atrial appendage, pulmonary arteries, pulmonary veins, ascending aorta, aortic
arch, superior vena cava (SVC), and inferior vena cava (IVC) for extremely realistic imaging - 46 in. L x 20 in. W x 12 in. H Sh. wt. 108 lb.
- What's in the Box?
- FAST Ultrasound Training Model
- Thermoform plastic storage shell with hook-and-loop straps
- Clear ultrasound refill solution (1-liter bottle)
- Blue ultrasound refill solution (235 mL bottle)
- Red ultrasound refill solution (235 mL bottle)
- Microfiber cleaning cloth
- Baby powder
- User guide CD
Specifications
Medical Assessment Model | |
Sonography for Trauma Assessment | |
Tan | |
FAST Training Model, Thermoform plastic storage shell with hook-and-loop straps |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction