Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Foerster Sponge Forceps, Sklar 9-1/2 in., Straight, Smooth, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 872095
Forceps, Foerster Sponge Forceps, Sklar, 9-1/2 in., Straight, Smooth, Stainless Steel, EA. Sklar Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster sponge forceps are often used in bariatric surgeries because of their longer length, and the curved version may be used for removing the placenta in OB/GYN procedures. This product is straight with oval, fenestrated, smooth tips and a length of 9-1/2 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-1763