Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Surgi-OR Foerster Sponge Forceps, Sklar 9-1/2 in., Curved, Serrated, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 95586
Forceps, Surgi-OR Foerster Sponge Forceps, Sklar, 9-1/2 in., Curved, Serrated, Stainless Steel, EA. Surgi-OR Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6546