Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOG2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FOG2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094876
View Documents
Novus Biologicals
NBP303827100UL
100 μg Each for $320.71
Add to cart
 
NB094877
View Documents
Novus Biologicals
NBP30382720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FOG2 Polyclonal antibody specifically detects FOG2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

FOG2
Unconjugated
Rabbit
Human, Mouse
transcription factor GATA4, modulator of, zinc finger protein 89B, zinc finger protein, multitype 2
Recombinant fusion protein containing a sequence corresponding to amino acids 982-1151 of human FOG2 (NP_036214.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
23414
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit