Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoklR Rabbit anti-Bacteria, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP266849
Description
FoklR Polyclonal antibody specifically detects FoklR in Bacteria samples. It is validated for Western Blot.Specifications
FoklR | |
1 mg/mL | |
Western Blot 1:1000-1:2000 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
Synthetic peptide within C terminal of Planomicrobium okeanokoites fokIR | |
Polyclonal | |
Immunogen affinity purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Affinity Purified | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Bacteria |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title