Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folic Acid, Powder, FCC, 95-102%, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
59-30-3
C19H19N7O6
441.40
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
59-30-3
C19H19N7O6
441.40
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1

F1079, 59-30-3, C19H19N7O6

Spectrum™ Folic Acid, Powder, FCC is a water soluble B vitamin used to fortify cold cereals, flour, breads, pasta, bakery items, cookies, and crackers. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.003
Lead (Pb) 2 mg/kg
Packaging Amber Glass Bottle
Percent Purity 95 to 102%
Water 0.085
Quantity 25 g