Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folic Acid, Powder, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
59-30-3
C19H19N7O6
441.40
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
59-30-3
C19H19N7O6
441.40
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1

FO105, 59-30-3, C19H19N7O6

Spectrum™ Folic Acid, Powder, USP is a water soluble B vitamin used to treat folic acid deficiency associated with ulcerative colitis, alcoholism, kidney dialysis, and liver disease and certain types of anemia. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Ignition Residue 0.003
Packaging Amber Glass Bottle
Percent Purity 97 to 102%
Quantity 25 g