Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Folliculin Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572548
Description
FLCN is located within the Smith-Magenis syndrome region on chromosome 17. Mutations in FLCN are associated with Birt-Hogg-Dube syndrome, which is characterized by fibrofolliculomas, renal tumors, lung cysts, and pneumothorax. Alternative splicing of FLCN results in two transcript variants encoding different isoforms.
Specifications
Folliculin | |
Polyclonal | |
Unconjugated | |
FLCN | |
Folliculin; BHD skin lesion fibrofolliculoma protein; Birt-Hogg-Dube syndrome protein; FLCN; BHD | |
Rabbit | |
Protein A | |
RUO | |
201163 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.75 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NFG4 | |
FLCN | |
KLH conjugated synthetic peptide between 505-534 amino acids from the C-terminal region of human FLCN | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction