Learn More
Follistatin Antibody (17-2), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP261941C
Catalog No. NBP261941C
Add to Cart
Description
Description
Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
Follistatin
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
FST
Recognizes human Follistatin isoform FS288, raised against recombinant human FS288
0.1 mL
Primary
This antibody recognizes human Follistatin isoform FS288.
Store at 4°C in the dark.
IgG1
ELISA, Immunohistochemistry, Western Blot
17-2
Western Blot, ELISA, Immunohistochemistry
follistatin, follistatin isoform FST317, FSActivin-binding protein
Mouse
Protein A purified
Cancer
10468
Human
Purified
Recommendations
Recommendations