Learn More
Follistatin Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0296-9D28, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212313AF350
Catalog No. NP212313A30
Add to cart
Description
Description
Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Follistatin
MM0296-9D28
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
follistatin, follistatin isoform FST317, FSActivin-binding protein
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FST
Human Follistatin
0.1 mL
Primary
10468