Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0296-9D28, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212313AF750

Catalog No. NP212313A70


Add to cart

Description

Description

Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Follistatin
MM0296-9D28
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
FST
Human Follistatin
Protein G purified
Cancer
Primary
10468.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
follistatin, follistatin isoform FST317, FSActivin-binding protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit