Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Mouse anti-Human, Clone: FST/4282, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Follistatin
Clone FST/4282
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 1 ug/ml, Immunocytochemistry
Applications Western Blot, ELISA, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122457
View Documents
Novus Biologicals
NBP30749210UG
100 μg
Each for $352.89
Add to cart
 
NB122458
View Documents
Novus Biologicals
NBP30749220UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifications

Specifications

Follistatin
0.2 mg/ml
Western Blot, ELISA, Immunocytochemistry
Purified
RUO
Human
follistatin, follistatin isoform FST317, FSActivin-binding protein
Recognizes human Follistatin isoform FS288, raised against recombinant human FS288.
Monoclonal
Protein A or G purified
FST/4282
Western Blot 1-2 ug/ml, ELISA 1 ug/ml, Immunocytochemistry
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
10468
IgG1
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit