Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Follistatin Mouse anti-Human, DyLight 350, Clone: 17-2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261941UV
Description
Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Follistatin | |
17-2 | |
Western Blot, ELISA, Immunohistochemistry | |
Purified | |
FST | |
Recognizes human Follistatin isoform FS288, raised against recombinant human FS288 | |
Protein A purified | |
Cancer | |
Primary | |
10468 | |
Human |
ELISA, Immunohistochemistry, Western Blot | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
follistatin, follistatin isoform FST317, FSActivin-binding protein | |
Mouse | |
IgG1 | |
0.1mL | |
Store at 4°C in the dark. | |
Monoclonal | |
This antibody recognizes human Follistatin isoform FS288. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title