Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Follistatin Antibody (H10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261940V
Description
Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Follistatin | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FST | |
Synthetic peptide corresponding to sequence CDEDQDYSFPISSILEW of the C- terminal region of human Follistatin 315 | |
0.1 mL | |
Primary | |
This antibody recognizes human Follistatin isoform FS315. | |
Store at 4°C in the dark. | |
IgG2a |
ELISA, Immunohistochemistry, Western Blot | |
H10 | |
Western Blot, ELISA, Immunohistochemistry | |
follistatin, follistatin isoform FST317, FSActivin-binding protein | |
Mouse | |
Protein A purified | |
Cancer | |
10468 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction