Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Mouse anti-Human, DyLight 488, Clone: 17-2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261941G

Catalog No. NBP261941G


Add to cart

Description

Description

Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

Follistatin
17-2
Western Blot, ELISA, Immunohistochemistry
50mM Sodium Borate with 0.05% Sodium Azide
10468
Mouse
IgG1
Primary
0.1mL
Store at 4°C in the dark.
This antibody recognizes human Follistatin isoform FS288.
ELISA,Immunohistochemistry,Western Blot
DyLight 488
Purified
follistatin, follistatin isoform FST317, FSActivin-binding protein
FST
Recognizes human Follistatin isoform FS288, raised against recombinant human FS288
Monoclonal
Protein A purified
Cancer
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit