Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin Mouse anti-Human, DyLight 594, Clone: H10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261940DL594

Catalog No. NB261940594


Add to cart

Description

Description

Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

Follistatin
H10
Western Blot, ELISA, Immunohistochemistry
Purified
FST
Synthetic peptide corresponding to sequence CDEDQDYSFPISSILEW of the C- terminal region of human Follistatin 315
Protein A purified
Cancer
Primary
10468
Human
ELISA, Immunohistochemistry, Western Blot
DyLight 594
PBS with 0.05% Sodium Azide
follistatin, follistatin isoform FST317, FSActivin-binding protein
Mouse
IgG2a
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human Follistatin isoform FS315.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit