Learn More
Follistatin Mouse anti-Human, DyLight 755, Clone: H10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261940IR
Catalog No. NBP261940IR
Add to cart
Description
Description
Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
Follistatin
H10
Western Blot, ELISA, Immunohistochemistry
Purified
FST
Synthetic peptide corresponding to sequence CDEDQDYSFPISSILEW of the C- terminal region of human Follistatin 315
Protein A purified
Cancer
Primary
10468
Human
ELISA, Immunohistochemistry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
follistatin, follistatin isoform FST317, FSActivin-binding protein
Mouse
IgG2a
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human Follistatin isoform FS315.