Learn More
Follistatin-related Gene Protein/FLRG Mouse anti-Human, Clone: 206714, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems MAB1288SP
 View more versions of this product
Catalog No. MAB1288SP
Add to cart
This item is not returnable. View return policy
Description
Description
Follistatin-related Gene Protein/FLRG/Fstl3 Monoclonal specifically detects Follistatin-related Gene Protein/FLRG/Fstl3 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
Follistatin-related Gene Protein/FLRG/Fstl3
Monoclonal
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O95633
FSTL3
Mouse myeloma cell line NS0-derived recombinant human Follistatin-related Gene Protein/FLRG Met27-Val263 Accession # O95633
25 μg
Primary
Detects human Follistatin-related Gene Protein/FLRG in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant mouse FLRG is observed.
Human
Purified
Western Blot, ELISA
206714
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FLRGFSRP, Follistatin-like 3, follistatin-like 3 (secreted glycoprotein), Follistatin-like protein 3, Follistatin-related gene protein, follistatin-related protein 3, FSTL3
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
10272
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1