Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Follistatin-related Gene Protein/FLRG Mouse anti-Human, Clone: 206714, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $475.00

Specifications

Antigen Follistatin-related Gene Protein/FLRG/Fstl3
Clone 206714
Immunogen Mouse myeloma cell line NS0-derived recombinant human Follistatin-related Gene Protein/FLRG Met27-Val263 Accession # O95633
Gene ID (Entrez) 10272
Gene Accession No. O95633
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1288SP
View Documents Promotion Details
R&D Systems
MAB1288SP
25 ug
Each for $119.60
Add to cart
 
MAB1288
View Documents Promotion Details
R&D Systems
MAB1288
500 ug
Each for $475.00
Add to cart
 
Description

Description

Follistatin-related Gene Protein/FLRG Monoclonal antibody MAB1288SP Specifically detects Follistatin-related Gene Protein/FLRG in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

Follistatin-related Gene Protein/FLRG/Fstl3
Mouse myeloma cell line NS0-derived recombinant human Follistatin-related Gene Protein/FLRG Met27-Val263 Accession # O95633
O95633
Reconstitute at 0.5 mg/mL in sterile PBS.
Primary
Detects human Follistatin-related Gene Protein/FLRG in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant mouse FLRG is observed.
Unconjugated
Purified
RUO
206714
10272
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Monoclonal
Mouse
IgG1
Protein A or G purified from hybridoma culture supernatant
Western Blot, ELISA
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit