Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR1 Mouse anti-Human, Unconjugated, Clone: LK26, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS217525UG
Description
FOLR1 Monoclonal antibody specifically detects FOLR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation.Specifications
FOLR1 | |
Monoclonal | |
Unconjugated | |
Folate receptor alpha;FR-alpha;Adult folate-binding protein;FBP;Folate receptor 1;Folate receptor;adult;KB cells FBP;Ovarian tumor-associated antigen MOv18 | |
Mouse | |
29.82 kDa calculated. | |
25 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
LK26 | |
NP_000793.1 | |
FOLR1, FOLR | |
Gestational choriocarcinoma cells (Luc-75(c)) | |
Protein G Purified | |
Apoptosis and Cancer | |
Clone LK26 is a mouse monoclonal antibody that detects human Folare receptor 1. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction