Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR1 Mouse anti-Human, Unconjugated, Clone: LK26, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen FOLR1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS21751MI
View Documents
MilliporeSigma
MABS2175100UG
100 μg
Each for $386.00
Add to cart
 
MABS21752MI
View Documents
MilliporeSigma
MABS217525UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

FOLR1 Monoclonal antibody specifically detects FOLR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation.
Specifications

Specifications

FOLR1
Unconjugated
Mouse
Folate receptor alpha;FR-alpha;Adult folate-binding protein;FBP;Folate receptor 1;Folate receptor;adult;KB cells FBP;Ovarian tumor-associated antigen MOv18
Gestational choriocarcinoma cells (Luc-75(c))
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin), Immunoprecipitation
Purified
Human
FOLR1, FOLR
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit