Learn More
FOLR2 Mouse anti-Human, Mouse (Negative), Alexa Fluor™ 750, Clone: EM-35, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262208AF750
Catalog No. NB002450
Add to cart
Description
Description
FOLR2 Monoclonal specifically detects FOLR2 in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
FOLR2
EM-35
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified, Conjugated
FOLR2
BW5147alpha,beta- cells
Protein A purified
Store at 4°C in the dark.
Monoclonal
Mouse monoclonal antibody EM-35 recognizes an extracellular epitope on FOLR2, a 30-40 kDa cell surface protein serving as a receptor for folic acid.
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 750
50mM Sodium Borate with 0.05% Sodium Azide
BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein
Mouse
IgG1
0.1 mL
Primary
2350
Human, Mouse (Negative)