Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR2 Rabbit anti-Human, Biotin, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289846B

Catalog No. NB109711


Add to cart

Description

Description

FOLR2 Monoclonal specifically detects FOLR2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FOLR2
1
PBS
BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein
This antibody was obtained from a rabbit immunized with purified, recombinant Human FOLR2 (P14207-1; Met1-His228).
Protein A purified
RUO
Primary
2350
Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit