Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR2 Recombinant Rat Monoclonal Antibody (CL10), Invitrogen™

Rat Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA547988

Encompass

Catalog No. PIMA547988


Add to Cart

Description

Description

FOLR2 Recombinant Monoclonal antibody specifically detects FOLR2 in Human samples. It is validated for Immunohistochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOLR2
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P01854
Rat
Affinity chromatography
RUO
3497
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG2a
Immunohistochemistry, Western Blot
CL10
Unconjugated
IGHE
BETA-HFR; FBP; FBP/PL-1; Fbp2; folate binding protein; folate binding protein 2; folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein
Polyclonal human IgE.
200 μg
Primary
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit