Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Lab-Aids™ Food Analysis, Limited Shelf Life Products for Science Education
Identify major nutrients in common foods, test common foods for presence of nutrients and quantitatively determine Vitamin C in foods.
Manufacturer:  Lab-Aids™ 6
Catalog No. S20665
Includes: 1 teacher's guide with MSDS; 10 student worksheets and guides; 2 plastic droppers; 20 slides; 10 test tubes; 1 drop control bottle of Fat Testing solution; 1 drop control bottle each of: Protein Testing solution Type A, Protein Testing solution Type B, Starch Testing solution, Sugar Testing solution, Vitamin C Standardizing solution, and Vitamin C Testing solution
Description
Description
Using simple and safe chemical tests, students can very quickly and easily identify the major nutrients in common foods. Students use step-by-step instructions to test common foods for the presence of carbohydrates, proteins, lipids. They also perform qualitative (detect a presence) and quantitative (how much) tests of Vitamin C by using a very simple drop-by-drop titration process. Over 100 tests for unknowns can be performed on each food group. This kit does not use corrosive agents or toxic substances. NOTE: Because of limited shelf-life of certain components, the Food Analysis Kit should be used within three weeks of receipt.
Order Info
Number of students: 10 to 40 Number of groups: Maximum of 10 groups
Specifications
Specifications
10-40 students
1 teacher's guide with MSDS; 10 student worksheets and guides; 2 plastic droppers; 20 slides; 10 test tubes; 1 drop control bottle of Fat Testing solution; 1 drop control bottle each of: Protein Testing solution Type A, Protein Testing solution Type B, Starch Testing solution, Sugar Testing solution, Vitamin C Standardizing solution, and Vitamin C Testing solution
Kit: Food Analysis (Limited Shelf Life)
Education