Learn More
Labconco™ FlaskScrubber™ Laboratory Glassware Washer Accessory, Pipet Inserts
SDPSCI_ED
For use with FlaskScrubber and FlaskScrubber Vantage Series Laboratory Glassware Washers.
Manufacturer:  Labconco™ 4494200/EMD
 View more versions of this product
Catalog No. S28358
Add to cart
Description
Description
  • Wash and dry pipets inside and out
  • Allows direct injection cleaning of pipets
  • Self-sealing silicone check valves allow washing of partial or full loads
  • May be placed in lower Spindle Rack in Labconco FlaskScrubber or FlaskScrubber Vantage Series
  • May fit up to 4 Inserts at once in FlaskScrubber or FlaskScrubber Vantage Series washers
Warranty and Services
1 year
Specifications
Specifications
Stainless Steel (304)
Holds 8 pipettes (1 - 10mL)
FlaskScrubber and FlaskScrubber Vantage Series Glassware Washers
2 lb.
Pipette Insert
Washing and drying pipets
One pipet holder and one mounting spindle
907 g